di-Natrium-L(+)-tartrat Dihydrat

Lieferant: Roth Carl

0254.1 0254.2 0254.3
ROTH0254.1EA 48.4 EUR
ROTH0254.1 ROTH0254.2 ROTH0254.3
di-Natrium-L(+)-tartrat Dihydrat
di-Natrium-L(+)-tartrat Dihydrat
Formel: NaOOC(CHOH)₂COONa·2H₂O
Schmelzpunkt: 57 °C
Dichte: 1,818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS-Nummer: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR