D(+)-Glucose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
M-Clarity™ quality level = MQ200
G5767-25G
G5767-500G
G5767-5KG
SIALG5767-25GEA
45.7
EUR
SIALG5767-25G
SIALG5767-500G
SIALG5767-5KG
D(+)-Glucose, Sigma-Aldrich®
D(+)-Glucose
Formel:
C₆H₁₂O₆ Schmelzpunkt: 146 °C Dichte: 1.54 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00148912 CAS-Nummer: 50-99-7 EINECS: 200-075-1 Merck Index: 14,04459 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...