Patentblau V Natriumsalz für die Mikroskopie C.I. 42051, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme: Food Blue 5 sodium salt, C.I. 42051, Acid blue 3 sodium salt, Bis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], sodium salt
21605-100G 21605-10G 21605-25G
21605-100GEA 267 EUR
21605-100G 21605-10G 21605-25G
Patentblau V Natriumsalz für die Mikroskopie C.I. 42051, Sigma-Aldrich®
Patentblau V Natriumsalz
Patent blue V sodium salt has been used as a stain in pharmacology. It is mainly used as a food color.
Synonyms: Acid blue 3 sodium salt; food blue 5 sodium salt
Formel: C₂₇H₃₁N₂NaO₇S₂
Molekulargewicht: 582.67
MDL Number: MFCD09608138
CAS-Nummer: 20262-76-4
EINECS: 243-654-4