D(+)-Saccharoseoctaacetat 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
419790010
419792500
419790051
ACRO419790010EA
126
EUR
ACRO419790010
ACRO419792500
ACRO419790051
D(+)-Saccharoseoctaacetat 98%
D(+)-Saccharoseoctaacetat
Formel:
C₂₈H₃₈O₁₉ Siedepunkt: 250 °C (1 mmHg) Schmelzpunkt: >78 °C Dichte: 1.28 g/cm³ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006623 CAS-Nummer: 126-14-7 EINECS: 204-772-1 Merck Index: 12,09052 |
Spezifikation Testergebnisse
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | ≥78 °C |
Water | ≤1 % (K.F.) |
Residue after ignition | ≤0.1 % |
Titration excess NaOH | 97.5 to 102.5 % (on anhydrous substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...