D(+)-Trehalose Dihydrat, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
625625-50
625625-2500
625625-1KG
625625-50EA
118
EUR
625625-50
625625-2500
625625-1KG
D(+)-Trehalose Dihydrat, Millipore®
D(+)-Trehalose Dihydrat
Formel:
C₁₂H₂₂O₁₁·2H₂O Molekulargewicht: 378.33 g/mol Schmelzpunkt: 97…99 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00071594 CAS-Nummer: 6138-23-4 EINECS: 202-739-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...