trans-Platin ≥64,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)
10472.01
10472.03
10472.06
10472.01EA
84.3
EUR
10472.01
10472.03
10472.06
trans-Platin ≥64,5%
trans-Platin
CAS No.: 14913-33-8
Molecular Formula: Cl2H6N2Pt
Formula Weight: 300.06
Physical Form: Crystalline
Appearance: Yellow
Odour: Odourless
MDL No.: MFCD00011623
Formel:
H₆Cl₂N₂Pt Schmelzpunkt: 340 °C (dec.) Dichte: 3.746 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011623 CAS-Nummer: 14913-33-8 EINECS: 238-980-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...