Cisplatin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride
M-Clarity™ quality level = MQ100
479306-5G
479306-1G
SIAL479306-5GEA
815
EUR
SIAL479306-5G
SIAL479306-1G
Cisplatin, Sigma-Aldrich®
cis-Platin
Formel:
H₆Cl₂N₂Pt Dichte: 3.738 g/cm³ (20 °C) |
MDL Number:
MFCD00011623 CAS-Nummer: 15663-27-1 EINECS: 239-733-8 UN-Nummer: 3288 ADR: 6.1,II Merck Index: 13,02341 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...