Paraffin, zähflüssig Reag. Ph. Eur.

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

1.07160.1000 1.07160.2500 1.07160.9026 1.07160.9181
1.07160.1000EA 69.9 EUR
1.07160.1000 1.07160.2500 1.07160.9026 1.07160.9181
Paraffin, zähflüssig Reag. Ph. Eur.
Paraffin
Dichte: 0,86 g/cm³ (20 °C)
CAS-Nummer: 8042-47-5
EINECS: 232-384-2
Merck Index: 13,07263

Bestellen

Spezifikation Testergebnisse

Identity (IR-spectrum) passes test
Identity (Viscosity) passes test
Acidity or alkalinity passes test
Relative Density (d 20/20) 0.827 - 0.890
Dynamic viscosity (20 °C) 110 - 230 mPa∙s
Aromatic, polycyclic Hydrocarbons passes test
Solid Paraffines passes test
Readily carbonisable substance passes test
Sulfur compounds passes test
Corresponds Ph Eur, BP, USP -


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR