Eisen ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 2.0 mm (0.08 in)

Lieferant: Thermo Fisher Scientific

10934.BU 10934.BY
10934.BUEA 381 EUR
10934.BU 10934.BY
Eisen ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 2.0 mm (0.08 in)
Iron

Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈25g/m MDL No.: MFCD00010999

Formel: Fe
Siedepunkt: 3000 °C (1013 hPa)
Schmelzpunkt: 1535 °C
Dichte: 7.86 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010999
CAS-Nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR