Zinn ≥99,998% (Metall-Basis), Drahtform, Puratronic®, Ø 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific

11907.G1 11907.G5 11907.G9
11907.G1EA 37.1 EUR
11907.G1 11907.G5 11907.G9
Zinn ≥99,998% (Metall-Basis), Drahtform, Puratronic®, Ø 0.25 mm (0.01 in)
Zinn

Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈0.36g/m Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Siedepunkt: 2270 °C (1013 hPa)
Schmelzpunkt: 232 °C
Dichte: 7.31 g/cm³ (25 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00133862
CAS-Nummer: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR