Gallium (Metall) ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

44418.22 44418.36
44418.22EA 690 EUR
44418.22 44418.36
Gallium (Metall) ≥99,999% (Metall-Basis)
Gallium

Packaged in poylethylene bottle.


Purity: 99.999% CAS No.: 7440-55-3 Physical Form: May be solid or liquid due to ambient temperature UN No.: 2803 MDL No.: MFCD00134045

Formel: Ga
Siedepunkt: 2403 °C (1013 hPa)
Schmelzpunkt: 29 °C
Dichte: 5.904 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00134045
CAS-Nummer: 7440-55-3
EINECS: 231-163-8
UN-Nummer: 2803
ADR: 8,III
Merck Index: 12,04364

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR