Gallium ≥99,99% (Metall-Basis), Barren
Lieferant: Thermo Fisher Scientific
|
Achtung
|
10188.18
10188.30
10188.14
10188.18EA
428
EUR
10188.18
10188.30
10188.14
Gallium ≥99,99% (Metall-Basis), Barren
Gallium
Purity: 99.99%
CAS No.: 7440-55-3
Molecular Formula: Requires overnight shipping with cold pack
Storage Temperature: 2°C to 8°C
Appearance: Silver grey
MDL No.: MFCD00134045
Formel:
Ga Siedepunkt: 2403 °C (1013 hPa) Schmelzpunkt: 29 °C Dichte: 5.904 g/cm³ (20 °C) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00134045 CAS-Nummer: 7440-55-3 EINECS: 231-163-8 UN-Nummer: 2803 ADR: 8,III Merck Index: 12,04364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...