Polyacrylic acid sodium salt, M.W. ~2100, Pulver, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
420344-100G
420344-500G
SIAL420344-100GEA
89.5
EUR
SIAL420344-100G
SIAL420344-500G
Polyacrylic acid sodium salt, M.W. ~2100, Pulver, Sigma-Aldrich®
Polyacrylic acid sodium salt
Formel:
[CH₂CH(CO₂Na)]n Dichte: 0,55 g/ml (25 °C) |
MDL Number:
MFCD00147949 CAS-Nummer: 9003-04-7 |
Spezifikation Testergebnisse
Infrared Spectrum | Conforms to Structure |
Water (by Karl Fischer) | 8 - 14% |
pH (C=1%, H₂O) | 6.0 - 9.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...