Anti-DNAJ Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADISPA410D ADISPA410F
ENZOADISPA410DEA 225 EUR
ENZOADISPA410D ENZOADISPA410F
Anti-DNAJ Rabbit Polyclonal Antibody
Antikörper
DnaJ is a basic 41 kDa E. coli Heat shock protein which belongs to the molecular chaperone class of proteins. Bacterial DnaJ protein acts synergistically with the bacterial chaperone DnaK (Hsp70 homologue) and the other co-chaperone GrpE in suppressing eukaryotic and prokaryotic polypeptide aggregation, facilitating protein translocation through intracellular compartments or protein secretion, reactivating some partially aggregated enzymes, and activating pre-priming complex during initiation of DNA replication.

  • For Immunoprecipitation, Western Blot

Recognizes bacteria DnaJ. Detects a band of ~41kDa by Western blot.

Type: Primary
Antigen: DNAJ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR