Anti-PKA Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADIKASPK017D ADIKASPK017F
ENZOADIKASPK017DEA 225 EUR
ENZOADIKASPK017D ENZOADIKASPK017F
Anti-PKA Rabbit Polyclonal Antibody
Antikörper
PKA (Protein Kinase A), also known as cAMP-dependent protein kinase, phosphorylates serine or threonine residues in target protein in response to elevated levels of cAMP. Inactive PKA exists as a tetrameric protein composed of two regulatory (R) subunits and two catalytic (C) subunits. Activation occurs when two cAMP molecules bind to each R subunit which causes a conformational change that releases the active C subunits.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus PKA. Detects a band of ~40kDa by Western blot.

Type: Primary
Antigen: PKA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR