Valacyclovir hydrochloride hydrate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride

462260050 462260250
ACRO462260050EA 125 EUR
ACRO462260050 ACRO462260250
Valacyclovir hydrochloride hydrate
Valacyclovir hydrochloride hydrate
Formel: C₁₃H₂₀N₆O₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01861507
CAS-Nummer: 124832-27-5

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Water 5.0 to 11.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR