Ternidazole 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Methyl-5-nitroimidazole-1-propanol

461950010 461952500
ACRO461950010EA 394 EUR
ACRO461950010 ACRO461952500
Ternidazole 97%
Ternidazole
Formel: C₇H₁₁N₃O₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01721389
CAS-Nummer: 1077-93-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR