Fenofibrat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate

461780050 461780250 461781000
ACRO461780050EA 54.5 EUR
ACRO461780050 ACRO461780250 ACRO461781000
Fenofibrat 98%
Fenofibrate
Formel: C₂₀H₂₁ClO₄
Schmelzpunkt: 79…82 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00133314
CAS-Nummer: 49562-28-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
Loss on drying ≤0.5 % (60 °C) (vacuum)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR