Terazosin hydrochloride dihydrate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Amino-6,7-dimethoxy-2-[4-(tetrahydro-2-furoyl)-1-piperazinyl]quinazoline Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(tetrahydrofuran-2-yl)methanone hydrochloride dihydrate

460860010 460860050
ACRO460860010EA 96 EUR
ACRO460860010 ACRO460860050
Terazosin hydrochloride dihydrate 98%
Terazosin hydrochloride dihydrate
Formel: C₁₉H₃₀ClN₅O₆
Schmelzpunkt: 271…274 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00941408
CAS-Nummer: 70024-40-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤9 % (105°C, 3 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR