Meloxicam 99-101%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
459550050
459550250
ACRO459550050EA
37.2
EUR
ACRO459550050
ACRO459550250
Meloxicam 99-101%
Meloxicam
Formel:
C₁₄H₁₃N₃O₄S₂ Schmelzpunkt: 255 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00868752 CAS-Nummer: 71125-38-7 |
Spezifikation Testergebnisse
Appearance | Light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C, 4 hr) |
Residue after ignition | ≤0.1 % |
Titration with HClO4 | 99.0 to 101.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...