Ertapenem sodium salt
Lieferant: Thermo Fisher Scientific
|
Achtung
|
460640010
460642500
ACRO460640010EA
0
EUR
ACRO460640010
ACRO460642500
Ertapenem sodium salt
Ertapenem sodium salt
Formel:
C₂₂H₂₄N₃NaO₇S Schmelzpunkt: 230…234 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD09475566 CAS-Nummer: 153773-82-1 |
Spezifikation Testergebnisse
Appearance | White to light yellow to light brown Powder |
Infrared spectrum | Conforms |
Loss on drying | 15 % to 19 % |
HPLC | ≥80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...