Bifonazol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(alpha,4-Diphenylbenzyl)imidazole, 1-([1,1'-Biphenyl]-4-yl(phenyl)methyl)-1H-imidazole
458620010
458620050
458620250
ACRO458620010EA
0
EUR
ACRO458620010
ACRO458620050
ACRO458620250
Bifonazol 98%
Bifonazole
Formel:
C₂₂H₁₈N₂ Schmelzpunkt: 149 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00865567 CAS-Nummer: 60628-96-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 148 to 153 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C, 1 h) |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...