4-Hydroxyphenyl benzoate 98%

Lieferant: Thermo Fisher Scientific
Achtung

455730050 455730250
ACRO455730050EA 41.6 EUR
ACRO455730050 ACRO455730250
4-Hydroxyphenyl benzoate 98%
4-Hydroxyphenyl benzoate
Formel: C₁₃H₁₀O₃
Molekulargewicht: 214.22 g/mol
Schmelzpunkt: 164…165 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00053304
CAS-Nummer: 2444-19-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 162 to 166 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR