Eisen(II)sulfat Heptahydrat ≥99%, bläulich grünes granulöses Pulver Reagenz für die Zellkultur
Lieferant: MP Biomedicals
Achtung
Synonyme: Iron vitriol, Ferrous sulphate heptahydrate
0219466383 0219466390
ICNA0219466383EA 61.1 EUR
ICNA0219466383 ICNA0219466390
Eisen(II)sulfat Heptahydrat ≥99%, bläulich grünes granulöses Pulver Reagenz für die Zellkultur
Eisen(II)sulfat Heptahydrat
Storage: Room Temperature, desiccate, store under nitrogen.
Ferrous sulfate heptahydrate is a reagent used in the manufacturing of iron compounds, in electroplating baths, aluminium etching, process engraving and lithography, and in redox polymerisation. It is used as a reducing agent in chemical processes, leather dyes and writing inks. Also, it is a component of weed killers, wood preservatives and water treatment processes. Used In prevention of chlorosis in plants and therapeutically as hematinic. In veterinary medicine, it is used as an astringent to combat iron deficiency. Ferrous sulfate heptahydrate is used in the determination of phosphorus as per the method of Taussky and Shorr. Ferrous sulfate heptahydrate is used in cell culture applications generally bound to transferrin, citrate or other iron transport molecules.
  • Cell Culture Reagent Grade
  • Auto Ignition: > 100 °C (Lit.)
  • Vapor Pressure: 14.6 mmHg ( 25 °C)
  • Presentation: Bluish Green Granular Powder
  • Soluble in water (100 mg/mL).
Formel: FeSO₄·7H₂O
Molekulargewicht: 278.02 g/mol
Siedepunkt: 2735 °C (1013 hPa) (Lit.)
Schmelzpunkt: 1539 °C (Lit.)
Dichte: 1.898 g/mL (25 °C) (Lit.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149719
CAS-Nummer: 7782-63-0
UN-Nummer: 3077
ADR: 9 III
Merck Index: 13,04091
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference