N-Boc-4-methylene-L-proline 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-1-(tert-Butoxycarbonyl)-4-methylenepyrrolidine-2-carboxylic acid

449260010 449260050
ACRO449260010EA 82.4 EUR
ACRO449260010 ACRO449260050
N-Boc-4-methylene-L-proline 97%
N-Boc-4-methylene-L-proline
Formel: C₁₁H₁₇NO₄
Schmelzpunkt: 110…115 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01861787
CAS-Nummer: 84348-38-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder or solid
Infrared spectrum Conforms
Melting point 110 to 115 °C
HPLC ≥96.0 %
Specific optical rotation -53° to -63° (20 °C, 589 nm) (c=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR