Flubendazol

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl (5-(4-fluorobenzoyl)-1H-benzo[d]imidazol-2-yl)carbamate, Methyl [5-(4-Fluorobenzoyl)benzimidazol-2-yl]carbamate, [5-(4-Fluorobenzoyl)benzimidazol-2-yl]carbamic acid Methyl Ester

101910010 101910050 101910250
ACRO101910010EA 21 EUR
ACRO101910010 ACRO101910050 ACRO101910250
Flubendazol
Flubendazole
Formel: C₁₆H₁₂FN₃O₃
Schmelzpunkt: 260 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00871999
CAS-Nummer: 31430-15-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 4 hr)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR