Fenofibrat
Lieferant: Cayman Chemical
Synonyme:
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
10005368-1
10005368-5
10005368-10
10005368-50
CAYM10005368-1EA
19
EUR
CAYM10005368-1
CAYM10005368-5
CAYM10005368-10
CAYM10005368-50
Fenofibrat
Fenofibrate
2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-propanoic acid, 1-methylethyl ester
Formel:
C₂₀H₂₁ClO₄ |
MDL Number:
MFCD00133314 CAS-Nummer: 49562-28-9 |
Spezifikation Testergebnisse
Formula weight | 360.8 |
Purity | ≥98% |
Formulation | Crystalline solid |
Storage temperature | -20°C |
Stability | ≥ 2 years |
λmax | 287 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...