2-Brom-1-chlor-4-iodbenzol 98%

Lieferant: Thermo Fisher Scientific
Achtung

458380010 458380050
ACRO458380010EA 89.2 EUR
ACRO458380010 ACRO458380050
2-Brom-1-chlor-4-iodbenzol 98%
2-Brom-1-chlor-4-iodbenzol
Formel: C₆H₃BrCl
Schmelzpunkt: 39…42 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07780667
CAS-Nummer: 31928-46-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder or crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 39 to 44 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR