5-Methylisatin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
206860010
206860050
ACRO206860010EA
36.4
EUR
ACRO206860010
ACRO206860050
5-Methylisatin 95%
5-Methylisatin
Formel:
C₉H₇NO₂ Schmelzpunkt: 180 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005721 CAS-Nummer: 608-05-9 |
Spezifikation Testergebnisse
Appearance | Orange-red to red-purple to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 186 to 190 °C |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...