Bottle-top dispensers, Varispenser® 2 and Varispenser® 2x
Lieferant: EPPENDORF
4966000.010 4966000.029 4966000.037 4966000.045 4966000.053 4966000.061 4967000.014 4967000.022 4967000.030 4967000.049 4967000.057 4967000.065
613-5915EA 325 EUR
613-5915 613-5916 613-5917 613-5918 613-5919 613-5920 613-5921 613-5922 613-5923 613-5924 613-5925 613-5926
Bottle-top dispensers, Varispenser® 2 and Varispenser® 2x
Flaschenaufsatz-Dispenser
Variable volume, autoclavable.
  • Discharge valve with safety ball closes and avoids fluid leakage when discharge tube is not mounted
  • Standard GL 45 thread on all sizes fits most common bottle threads
  • Screwable air vent cap allows easy assembly of optional drying tube to protect sensitive reagent against humidity or CO₂
  • Oval shaped dispenser housing allows safe gripping for transportation and adjustment
For laboratory tasks with aliquots of aggressive solutions like lyes, acids, bases or solvents taken from large supply and reagent bottles, bottle top dispensers need to be universally applicable and of uncompromising quality.
The Varispenser 2 and Varispenser 2x bottle top dispensers offer high chemical resistance and robustness in the range of volumes up to 100 ml. Varispenser 2x is additionally equipped with a recirculation valve for priming the dispensing system without loss of reagent.
The trusted PFA sealing lip technology prevents crystal building from media residue by wiping off the inner cylinder wall during dispensing. This technology combined with the high chemical resistance allows the use with nearly all kinds of liquids. The extra air vent cap with Luer-cone for the assembly of a microfilter prevents possible contamination from the supply bottle.
Varispenser 2 and Varispenser 2x are completely autoclavable. The 360° rotating valve block offers safe and easy access in all positions. Because of the GL 45 thread on all sizes, Varispenser 2 and Varispenser 2x can be used on most common bottle threads in the laboratory.
Zertifizierungen: EN ISO 8655
Bestellinformation: Supplied with a telescopic filling tube, wrench and five adapters: GL 25, GL 28, GL 32, GL 38 and S 40/buttress thread (for the three smaller Varispenser® 2/ Varispenser® 2x models) or with a filling tube, wrench and three adapters: GL 32, GL 38 and S 40/buttress thread (for the 3 larger Varispenser® 2/ Varispenser® 2x models).