Wasser RNase frei

Lieferant: Abnova

R0013-3 R0013-2
ABNOR0013-3EA 171 EUR
ABNOR0013-3 ABNOR0013-2
Wasser RNase frei
Wasser

RNase-free water are prepared from highly purified water by ultrafiltration without DEPC treatment.


  • Free from RNase and DNase
  • Ideal in RNA research

Formel: H₂O
MDL Number: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR