Cäsiumchlorid ≥99,0%, weißes kristallines Pulver, Reagenzqualität

Lieferant: MP Biomedicals
Achtung

Synonyme: Cesium Chloride

0210132191 0210132125 0210132180 0210132183 0210132190
ICNA0210132191EA 1350 EUR
ICNA0210132191 ICNA0210132125 ICNA0210132180 ICNA0210132183 ICNA0210132190
Cäsiumchlorid ≥99,0%, weißes kristallines Pulver, Reagenzqualität
Cesium chloride

Cesium chloride, an inorganic compound, is an important source of cesium ions in a variety of applications. Its crystal structure forms a major structural type where each cesium ion is coordinated by 8 chlorine ions. Cesium chloride occurs naturally in mineral waters and as an impurity in carnallite (up to 0.002%), sylvite and kainite. Less than 20 tonnes of CsCl is produced annually worldwide, mostly from a cesium-bearing mineral pollucite.


  • Reagent Grade
  • Refractive Index: 1.642(Lit.)
  • UV/Visible Absorbance: A50%/280, H2O ≤0.2(Lit.)
  • Presentation: White Powder
  • Storage temperature: Store at Room Temperature (15-30 °C), desiccate.


Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins. It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation. Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA. Procedures have been reported for the purification of closed circular DNA by equilibrium centrifugation in both continuous and discontinuous CsCl-ethidium bromide gradients. Subsequently, the CsCl can be removed from the DNA preparation by either dialysis or precipitation.


pH: 5.5 - 6.5 (1% aq soln)

Formel: CsCl
Molekulargewicht: 168.36 g/mol
Siedepunkt: 1303 °C (Lit.)
Schmelzpunkt: 646 °C (Lit.)
Dichte: 3.99 g/mL (Lit.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010955
CAS-Nummer: 7647-17-8
Merck Index: 13,02021

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99.0%
pH (1% aq soln) 5.5-6.5
Solubility (50% aq soln) Clear and Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR