C-Phycocyanin
Lieferant: Merck
C-Phycocyanin (CPC) is a pigment-protein complex from the light-harvesting phycobiliprotein family, it can be used in immunoassay kit development and to study its properties as a light harvesting protein.
- Storage temperature ranges from 2 to 8 °C
- Concentration: ≥10,0 mg/ml (UV)
- Fluorescence: λex 609; λem 643 nm in 0,1 M phosphate (pH 7,2)
This is suspended in 150 mM sodium phosphate, 60% ammonium sulphate, 1 mM EDTA, 1 mM sodium azide, pH 7,0 and must be dialysed against conjugation buffer or PBS before conjugation.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...