Containers for Sterilisation
Lieferant: MUEHLACKER METALWARENFABRIK
01574 01568 01564 01570 01576
MUEH01574EA 890 EUR
MUEH01574 MUEH01568 MUEH01564 MUEH01570 MUEH01576
Containers for Sterilisation
Boxen Transportboxen
Sterilisation containers are made of 18/10 stainless steel.
  • With seamless bottom or floor insert
  • Satin finish brushed
  • Close the hole with the cover
  • Bucket handle made of flat steel
  • Liquid tightness
Stackable seamless bucket allows for a simple and thorough cleaning by the great radius on the ground.