Plug valves, Parker Balston® shut-off gas valve
Lieferant: Restek
22196 22197 22198 22199
REST22196EA 0 EUR
REST22196 REST22197 REST22198 REST22199
Plug valves, Parker Balston® shut-off gas valve
Ventile Kegelventile
These shut-off plug valves work well in any application which requiring throttling or on/off operations.
  • Leak free at wide temperature swings
  • Rated to 3000 psig at 21 °C
  • Maximum operating temperature of 205 °C