Buffer solutions as per DIN 19266-01: 2000
Lieferant: WTW
109110
109120
109130
109400
109000
109020
WTWA109110EA
30.3
EUR
WTWA109110
662-0326
662-0327
662-0333
662-0332
WTWA109020
Buffer solutions as per DIN 19266-01: 2000
Puffer
pH-Puffer
These standard buffer solutions (work reference buffer solutions) are widely used in precision applications.
- Traceably certified to PTB/NIST and therefore safe for use
- Every bottle is labeled with their own lot number with an expiration date
Zertifizierungen: DIN 19266-01: 2000.