GC columns, Mxt®-Alumina BOND/MAPD
Lieferant: Restek
79728 79728-273
REST79728EA 1507 EUR
REST79728 REST79728-273
GC columns, Mxt®-Alumina BOND/MAPD
Chromatographiesäulen
MXT-Alumina BOND/MAPD PLOT columns are made specifically for the analysis of petrochemicals and downstream products such as ethylene, propylene, butylenes and butadiene.
  • Optimised deactivation produces maximum response when analysing trace levels of acetylene, methyl acetylene and propadiene
  • High loadability reduces peak tailing and improves separations
  • Extended temperature range up to 250 °C for fast elution of high molecular weight (HMW) hydrocarbons and accelerated column regeneration following exposure to water
  • Metal Siltek-treated tubing is incredibly durable, allowing MXT columns to be tightly coiled without breaking for use in smaller GC ovens
Alumina PLOT deactivation produces an incredibly inert column that provides superior reproducibility and stable response factors to maximise the number of analyses before recalibration is required.