Column kits, Rtx®-CLPesticides
Lieferant: Restek
11197 11199
REST11197EA 1843 EUR
REST11197 REST11199
Column kits, Rtx®-CLPesticides
Chromatographiesäulen
These Rtx-CLPesticides column kits are suitable for organochlorine pesticides and herbicides. Analyse EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1 and 552.2 compounds without time consuming column changes.
  • Baseline separations in less than 10 minutes
  • Temperature range: stable from 320 to 340 °C
  • Low bleed, ideal for high sensitivity GC-ECD or GC-MS analyses
Note: Analysing dirt or derivatised samples can contaminate the column. Not recommended for analysing trace-level pesticide samples following derivatised samples (e.g., Methods 8151A and 552.2) without first performing inlet maintenance. Standard steps include trimming the guard column and changing the inlet liner, O-ring, seal and septum.
Lieferumfang: Kit contains an Rtx-CLPesticides column, Rtx-CLPesticides2 column, deactivated universal angled 'Y' press-tight connector and IP deactivated guard column. Columns are not preconnected in this kit.