Anti-AR Mouse Monoclonal Antibody (CF568) [clone: AR441 DHTR/882]

Lieferant: Biotium
BNC680913-500 BNC680913-100
BNC680913-500EA 560 EUR
BNC680913-500 BNC680913-100
Anti-AR Mouse Monoclonal Antibody (CF568) [clone: AR441 DHTR/882]
Antikörper
Recognizes a protein of 110 kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy. Anti-androgen receptor has been useful clinically in differentiating morpheaform basal cell carcinoma (mBCC) from desmoplastic trichoepithelioma (DTE) in the skin.This MAb is superb for staining of formalin/paraffin tissues.

Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: AR441 DHTR/882
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR