Anti-KRT7 Mouse Monoclonal Antibody (CF647) [clone: KRT7/760 KRT7/903]

Lieferant: Biotium
BNC470906-500 BNC470906-100
BNC470906-500EA 560 EUR
BNC470906-500 BNC470906-100
Anti-KRT7 Mouse Monoclonal Antibody (CF647) [clone: KRT7/760 KRT7/903]
Antikörper
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/760 KRT7/903
Conjugation: CF647
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgGs
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR