Anti-AR Mouse Monoclonal Antibody (Purified) [clone: AR441]

Lieferant: Biotium
BNUB0881-500 BNUB0881-100
BNUB0881-500EA 511 EUR
BNUB0881-500 BNUB0881-100
Anti-AR Mouse Monoclonal Antibody (Purified) [clone: AR441]
Antikörper
Recognizes a protein of 110 kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy. Anti-androgen receptor has been useful clinically in differentiating morpheaform basal cell carcinoma (mBCC) from desmoplastic trichoepithelioma (DTE) in the skin.This MAb is superb for staining of formalin/paraffin tissues.

Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: AR441
Conjugation: Purified
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR