Anti-KLK3 Mouse Monoclonal Antibody (Biotin) [clone: KLK3/801]

Lieferant: Biotium
BNCB0801-500 BNCB0801-100
BNCB0801-500EA 560 EUR
BNCB0801-500 BNCB0801-100
Anti-KLK3 Mouse Monoclonal Antibody (Biotin) [clone: KLK3/801]
Antikörper
Recognizes a single protein of 33-34 kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: KLK3/801
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR