Anti-PRLR Mouse Monoclonal Antibody (CF594) [clone: B6.2 PRLR742]

Lieferant: Biotium
BNC940744-500 BNC940744-100
BNC940744-500EA 560 EUR
BNC940744-500 BNC940744-100
Anti-PRLR Mouse Monoclonal Antibody (CF594) [clone: B6.2 PRLR742]
Antikörper
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: B6.2 PRLR742
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR