Bronopol, weißes kristallines Pulver

Lieferant: MP Biomedicals
Gefahr

Synonyme: 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol

0215488725 0215488780
ICNA0215488725EA 57.7 EUR
ICNA0215488725 ICNA0215488780
Bronopol, weißes kristallines Pulver
Bronopol

Soluble in a 1% aqueous solution (clear, colorless solution).


Soluble in a 1% aqueous solution (clear, colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: HOCH₂BrC(NO₂)CH₂OH
Schmelzpunkt: 125…135 °C
MDL Number: MFCD00007390
CAS-Nummer: 52-51-7
UN-Nummer: 3241
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR