Polyethylenglycol 8.000, Pulver
Lieferant: MP Biomedicals
Synonyme:
PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water.
PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridisation. PEG 8 000, can be used to precipitate DNA. PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis.
PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds. PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives
Soluble in aqueous solution (50 mg/ml - clear, colorless solution).
Formel:
H(OCH₂CH₂)nOH Molekulargewicht: 8000 g/mol Siedepunkt: 250 |
MDL Number:
MFCD01779615 CAS-Nummer: 25322-68-3 Merck Index: 12,07729 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 56 - 66 °C |
pH (5% aq soln) | 5.0 - 7.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...