Nalidixinsäure Natriumsalz
Lieferant: MP Biomedicals
Synonyme:
Sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Nalidixic acid sodium salt is an antibacterial agent (synthetic quinolone antibiotic), which is inhibitor of bacterial DNA polymerase (DNA gyrase) & avian myeloblastoma virus reverse transcriptase.
Nalidixic acid sodium salt is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. Inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.
Typically used at a concentration of 15 ug/ml in molecular biology.
Soluble in water (100 mg/ml - clear, light yellow solution). Stock solutions can be aliquoted and stored at -20 °C for up to approximately 3 to 4 months.
Formel:
C₁₂H₁₁N₂NaO₃ Molekulargewicht: 254.22 g/mol |
MDL Number:
MFCD00064376 CAS-Nummer: 3374-05-8 |
Spezifikation Testergebnisse
Form | Off-white powder |
Storage | Store at 0 °C, Use only in area provided with appropriate exhaust ventilation |
Moisture Content | ≤9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...