Bromthymolblau Natriumsalz, Pulver

Lieferant: MP Biomedicals

Synonyme: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

0215052405 0215052425
ICNA0215052405EA 35.9 EUR
ICNA0215052405 ICNA0215052425
Bromthymolblau Natriumsalz, Pulver
Bromthymolblau Natriumsalz
Formel: C₂₇H₂₇Br₂NaO₅S
Molekulargewicht: 646.37 g/mol
MDL Number: MFCD00077263
CAS-Nummer: 34722-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR