Aztreonam, weißes Pulver
Lieferant: MP Biomedicals
Synonyme:
2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
Aztreonam is the first totally synthetic monocyclic β-lactam antibiotic that displays a high degree of resistance to β-lactamase degradation. It has high specificity for gram-negative aerobic rods.
Aztreonam specifically targets the penicillin binding protein 3 (PBP-3) of Gram-negative bacteria. It is also effective against such Enterobacteriaceae as E. coli, Klebsiella, Proteus, Providencia, Salmonella, Serratia, Shigella, and Yersinia species.
Soluble in DMF and DMSO; slightly soluble in ethanol and methanol; practically insoluble in toluene, chloroform and ethyl acetate.
Bitte beachten Research Use Only (RUO).
Formel:
C₁₃H₁₇N₅O₈S₂ Schmelzpunkt: 227 °C (dec., lit.) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00072145 CAS-Nummer: 78110-38-0 |
Spezifikation Testergebnisse
Identity Test | Passes |
Potency (as is) | Report Result (µg/mg) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...