HEPES Natriumsalz ≥98,0%, kristallin

Lieferant: MP Biomedicals

0210559325 0210559380 0210559383 0210559391
ICNA0210559325EA 51.6 EUR
ICNA0210559325 ICNA0210559380 ICNA0210559383 ICNA0210559391
HEPES Natriumsalz ≥98,0%, kristallin
HEPES Natriumsalz
Formel: C₈H₁₇N₂NaO₄S
MDL Number: MFCD00036463
CAS-Nummer: 75277-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Synonyms 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt; N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulphonic acid) sodium salt
Form White Powder
Storage Store at Room Temperature (15 - 30 °C), Desiccate
Loss on Drying 3,0%
pH 9,5 - 10,8 (1% aq soln)
pka 7,55 at 25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR