DL-Serin ≥98%, weißes Pulver
Lieferant: MP Biomedicals
Synonyme:
(±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid
0210286825
0210286880
0210286891
ICNA0210286825EA
57.8
EUR
ICNA0210286825
ICNA0210286880
ICNA0210286891
DL-Serin ≥98%, weißes Pulver
DL-Serin
NMDA agonist acting at the glycine site; precursor of glycine by serine hydroxymethyltransferase.
Soluble in water (50 mg/ml - clear colorless solution).
Bitte beachten Research Use Only (RUO).
Formel:
C₃H₇NO₃ Schmelzpunkt: 228 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064223 CAS-Nummer: 302-84-1 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Melting Point | 234 - 244 °C |
Optical Rotation (c = 5, 1N HCl) | -1° to + 1° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...